Learn More
Fisherbrand™ Aluminum Lab Frame Rods
Product Code. 17291384 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (English):
12 in.
Length (Metric):
30.48 cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Length (Metric) Unit Size
17291384 12 in. 30.48 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17291384
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Spedizione stimata: 29-12-2026
Corrosion resistant hard aluminum rods for smooth fit with lab frames.
  • Rods with center-less ground allow for precise fiting
  • With chamfered ends for easy location
  • Diameter: 13mm (0.51 in.)
Specifications
Material Aluminum
Length (English) 12 in.
Length (Metric) 30.48 cm
Product Type Corrosion resistant with chamfered ends for easy location