Learn More
Hellma Cylindrical Cells
Product Code. 12711705
Change view
Click to view available options
Path Length:
10 mm
50 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Path Length Unit Size
12711705 10 mm Each
10126690 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12711705 Supplier Hellma Supplier No. 1201040
  • ONLINE EXCLUSIVE PRICE
    165.00€
    151.00€ / pezzi 1
    Risparmia 14.00€
    8% Sconto
Spedizione stimata: 04-12-2026
Fit standard cell holders
Cylindrical cells have a filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).
  • Uniform diameter of 22mm
  • Available with Suprasil™ quartz or near-UV glass windows
Specifications
Material Suprasil
Includes Suprasil quartz window; One stopper
Path Length 10 mm