Learn More
Fisherbrand™ Lid for 96/384 Well Polystyrene Plates
Product Code. 11927964 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Plate Compatibility:
96-well Plate
Unit Size:
Pack of 180
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plate Compatibility Unit Size
11927964 96-well Plate Pack of 180
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11927964
Spedizione stimata: 14-01-2027
Add to basket
Add to basket
Bulk Quote Request
Protect samples from contamination and evaporation during assay processing, incubation or storage
  • Clear
  • Non-sterile
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polystyrene
Sterility Non-sterile
No. per Case 180
For Use With (Application) Ideal for low-volume assays such as colorimetric, enzymatic, cloning, replication, array and storage
Product Type Plate Lid
Plate Compatibility 96-well Plate