Learn More
Fisherbrand™ Autoclavable HD Biohazard Waste Bags
Codice prodotto. 18356513 Sfoglia Tutto Fisherbrand Prodotti
Cambia vista
Click to view available options
Colore:
Clear with Red Print
Red with Black Print
Dimensioni (L x P):
12 x 8 in. (30 x 20 cm)
23 x 19 in. (58 x 48 cm)
24 x 12 in. (61 x 30 cm)
30 x 24 in. (76 x 61 cm)
36 x 24 in. (91 x 61 cm)
Spessore:
1.25 mil
1.50 mil
Dimensione della confezione:
pezzi 100
pezzi 1000
pezzi 200
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Colore Dimensioni (L x P) Spessore unitSize
18356513 Clear with Red Print 30 x 24 in. (76 x 61 cm) 1.50 mil pezzi 100
18568056 Clear with Red Print 24 x 12 in. (61 x 30 cm) 1.50 mil pezzi 100
18588056 Red with Black Print 12 x 8 in. (30 x 20 cm) 1.25 mil pezzi 1000
18376533 Clear with Red Print 36 x 24 in. (91 x 61 cm) 1.50 mil pezzi 100
18336513 Red with Black Print 23 x 19 in. (58 x 48 cm) 1.25 mil pezzi 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Codice prodotto. 18356513
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Strong and leak-resistant
  • Printed with biohazard symbol and text in English and Spanish
  • Supplied with twist tie closure
Specifica
Tipo di chiusura Twist Tie
Colore Clear with Red Print
Dimensioni (L x P) 30 x 24 in. (76 x 61 cm)
Lunghezza (sistema metrico) 76 cm
Larghezza (sistema metrico) 61 cm
Autoclavabile Autoclavable
Include Biohazard symbol and text in English and Spanish
Larghezza (sistema imperiale) 24 in.
Lunghezza (sistema imperiale) 30 in.
Spessore 1.50 mil
Materiale Polyethylene