Learn More
Fisherbrand™ Polypropylene Clamp with Flow Control
Regulates flow through tubing from fully open to closed.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.04500kg
Descrizione
- Fits 0.12 to 0.5 in. (3.2 to 13mm) O.D. tubing
- Molded white polypropylene construction
- Autoclavable
- With 15-position control
- Will not distort, rust, or corrode
Specifica
Autoclavable | |
Polypropylene | |
1.5 in. | |
Medium |
Tubing Clamp | |
Tube with 0.12 to 0.5 in. (0.304 to 1.27cm) OD | |
3.81 cm | |
White |