missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems FITC-Labeled Human CLEC12A / MICL / CLL-1 Protein, His Tag
444.19€ - 3017.85€
Specifica
Numero di accesso | NP_612210.4 |
---|---|
Peso molecolare | 25.6 kDa |
Sorgente | FITC-Labeled Human CLEC12A, His Tag (CLA-HF247) is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2). It is the FITC labeled form of Human CLEC12A, His Tag (CLA-H5245). |
Immunogeno | His 65 - Ala 265 |
Requisiti di stoccaggio | -20°C, avoid light |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17116261
|
ACROBiosystems
CLA-HF247-25UG |
25 μg |
444.19€
25µg |
Spedizione stimata: 29-04-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
17106261
|
ACROBiosystems
CLA-HF247-200UG |
200 μg |
3017.85€
200µg |
Spedizione stimata: 29-04-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
CLEC12A (C-type lectin domain family 12 member A) is also known as CLL1, DCAL2, MICL. Clec12a is an inhibitory receptor for uric acid crystals that regulates inflammation in response to cell death. Cell surface receptor that modulates signaling cascades and mediates tyrosine phosphorylation of target MAP kinases. Evidence of distinct disease propagating stem cells in myelodysplastic syndrome (MDS) has emerged in recent years. The role of CLEC12A in MDS, however, remains to be elucidated. Furthermore, CLEC12A has been proposed as a promising marker of leukaemic stem cells in AML.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 25.6 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
NP_612210.4 | |
FITC-Labeled Human CLEC12A, His Tag (CLA-HF247) is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2). It is the FITC labeled form of Human CLEC12A, His Tag (CLA-H5245). | |
-20°C, avoid light | |
CLEC12A | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |
25.6 kDa | |
His 65 - Ala 265 | |
CLEC12A,MICL,CLL-1,CLL1,DCAL2,DCAL-2,CD371 | |
FITC | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |