Learn More
Thermo Scientific™ Fixed-Needle, Gas-Tight GC Syringe, Pk. Of 6
Product Code. 15284563 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15284563 10 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15284563 Supplier Thermo Scientific™ Supplier No. 365D0626
Spedizione stimata: 30-12-2026
Thermo Scientific™ syringes for GC instruments are designed to provide enhanced durability,clarity and accuracy in sample introduction.
TRUSTED_SUSTAINABILITY
Specifications
Needle Gauge 23 to 26 G
Product Type Gas Tight GC Autosampler Syringe, Fixed Needle
Volume (Metric) 10 μL
Length (Metric) Needle 42 mm
Needle Point Style Cone
For Use With (Equipment) Agilent GC Instruments