missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress FLAG peptide

Codice prodotto. 16472689
Encompass_Preferred
Cambia vista
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16472689 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16472689 Fornitore MedChemExpress N. del fornitore HYP0223

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

MedChemExpress FLAG peptide is an eight amino acids peptide (Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys) with an enterokinase-cleavage site; designed for antibody-mediated identification and purification of recombinant proteins.

Specifications

Purezza 98.18%
CAS 98849-88-8
Content And Storage Protect from light, stored under nitrogen∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
Da utilizzare con (applicazione) COVID-19-immunoregulation
Peso molecolare 1012.97 g/mol
Product Type Peptides
Quantità 1 mg
Shipping Condition Room temperature
Solubilità H2O : 100 mg/mL (98.72 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.