Learn More
Thermo Scientific™ Flat Caps for UTW PCR Plates, strips of 8, optical clear
Product Code. 10128379 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
120 Strips
Unit Size:
Pack of 120
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10128379 120 Strips Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10128379 Supplier Thermo Scientific™ Supplier No. TCS1080
Black Friday Starts Here
Use promo code 27984
View all promotions
In stock
Add to basket
Edge
Add to basket
Bulk Quote Request
Minimize vapor loss with these flat cap strips for use with Thermo Scientific™ Piko™ 24-well plates in the Piko Thermal Cycler and the PikoReal Real-Time PCR System.
All our sealing products are designed to provide the ultimate protection while also being easy to use. Thermo Scientific qPCR sealing options are optically clear to deliver maximum and consistent signal transmission, critical for accurate qPCR results.
These flat cap strips provide an extremely tight seal in UTW (ultra-thin walled) PCR plates. They are an ideal choice for low volume reactions where it is important to minimize vapor loss. In addition, our optically-clear flat caps provide great transmission for all fluorescence detection applications, such as qPCR.
Highlights
  • 8-cap strip
  • Optically clear for fluorescence detection
  • Tight sealing for low-volume samples and consistent quantification results
  • Ideal for sealing 24-well Piko UTW PCR plates
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polypropylene
No. of Wells 8
Quantity 120 Strips
Format Strips of 8 Optical Caps
Product Type PCR Plate Cap Strip
Product Line Piko
Certified RNase/DNase-free Yes
Plate Compatibility 96-well Plate