Learn More
Flavianic Acid, TRC
Product Code. 30009627
Change view
Click to view available options
Quantità:
1 g
2.5 g
Unit Size:
1g
2.50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
30009627 1 g 1g
30005296 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30009627 Supplier Toronto Research Chemicals Supplier No. TRCF2573951G
 Articolo soggetto a restrizioni
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 483-84-1
Formula molecolare C10H6N2O8S
Molecular Weight (g/mol) 314.23
SMILES Oc1c(cc(c2ccc(cc12)S(=O)(=O)O)[N+](=O)[O-])[N+](=O)[O-]
Specifications
Materiale o nome chimico Flavianic Acid
Quantità 1 g
InChI Formula InChI=1S/C10H6N2O8S/c13-10-7-3-5(21(18,19)20)1-2-6(7)8(11(14)15)4-9(10)12(16)17/h1-4,13H,(H,18,19,20)
Peso formulazione 313.9845