Learn More
Kimble Chase™ Flex-Column™ Borosilicate Glass Economy Column
Product Code. 11742354
Change view
Click to view available options
Length (Metric):
500 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11742354 500 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11742354 Supplier Kimble Chase™ Supplier No. 4204000750
  • ONLINE EXCLUSIVE PRICE
    349.00€
    287.58€ / pezzi 10
    Risparmia 61.42€
    17% Sconto
 Restricted Item
Spedizione stimata: 08-02-2027
Specifications
USP Type Type I
Max. Pressure 1 bar
Column Capacity 20 mL
Length (Metric) 500 mm
Material Borosilicate Glass
Cross Sectional Area 0.39cm3
Temperature (Metric) Range 0 to 50°C
Quantity 10 Pack
For Use With (Application) Ion exchange chromatography
Type Economy Chromatography Column
Diameter (Metric) Inner 7 mm