Learn More
Corning™ Flex Discharge Tubes
Product Code. 16872120 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2.5 to 10 mL
25 to 100 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
16872120 25 to 100 mL Each
16862120 2.5 to 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16872120 Supplier Corning™ Supplier No. 6878
  • ONLINE EXCLUSIVE PRICE
    29.10€
    27.16€ / pezzi 1
    Risparmia 1.94€
    6% Sconto
Spedizione stimata: 09-04-2027
Corning™ Flex Discharge Tubes for quick and precise dispensing of acids, bases and solvents
  • Coiled
  • Includes protective sleeve
TRUSTED_SUSTAINABILITY
Specifications
Product Type Flexible Discharge Tube
Description Coiled
Material FEP, Polypropylene
Volume (Metric) 25 to 100 mL
For Use With (Application) Quick and precise dispensing of acids, bases and solvents
For Use With (Equipment) 25 to 100 mL Bottle Top Dispenser
Includes Protective Sleeve