Learn More
Molecular Devices FlexStation™ 3 Microplate Reader without Pipettor Head
Sfoglia Tutto Molecular Devices ProdottiDescrizione
- Designed for multi-mode detection workflows in microplate-based assays
- Supports biochemical and cell-based assay applications
- Enables flexible assay setup with configurable detection parameters
- Supports microplate-based workflows across multiple life science applications
- Designed for integration with automated liquid handling using optional pipettor head
- Integrated with SoftMax Pro Software for data acquisition and analysis
- Supports scalable assay workflows for research environments
Specifica
Specifica
| Product Type | Microplate Reader |
| Tipo di rilevatore | Photomultiplier Tube (PMT) |
| Interfaccia | PC Control, Internal Software |
| Sorgente luminosa | Xenon Flash Lamp |
| Tecnologia di misurazione | Fluorescence Intensity (Top and Bottom), Luminescence (Top), Absorbance (Top) |
| Tipo di piastra | 96 and 384-well Plates |
| Reading Options | Endpoint, Kinetic, Spectra |
| Agitazione | Linear |
| Selezione della lunghezza d'onda | Monochromator |