missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FLJ11127 Recombinant Protein
Marca: Abnova™ H00054491-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifica
AAH11524 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
64.9 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MAATRSPTRARERERSGAPAAGSDQVHSWMLATSQALDTVWRMAKGFVMLAVSFLVAAICYFRRLHLYSGHKLKWWIGYLQRKFKRNLSVEAEVDLLSYCAREWKGETPRNKLMRKAYEELFWRHHIKCVRQVRRDNYDALRSVLFQIFSQGISFPSWMKEKDIVKLPEKLLFSQGCNWIQQYSFGPEKYTGSNVFGKLRKYVELLKTQWTEFNGIRDYHKRGSMCNTLFSDAILEYKLYEALKFIMLYQVTEVYEQMKTKKVIPSLFRLLFSRETSSDPLSFMMNHLNSVGDTCGLEQIDMFILGYSLEVKIKVFRLFKFNSRDFEVCYPEEPLRDWPEISLLTENDRHYHIPVF | |
FLJ11127/FLJ43660 | |
FAM105A | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
54491 | |
FLJ11127 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FAM105A | |
Human | |
Recombinant | |
Solution |