Learn More
Thermo Scientific™ Fluid Transfer Assembly, Adaptor Set
Codice prodotto. 17850577 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Tipi di connettore:
Luer to MPC
MPC/MPX
MPX to MPC
MPX/MPC
Dimensioni dei tubi:
C-Flex 374, 2 m (6.6 ft)
C-Flex 374, 2 m (78.7 in)
TPE 374, 1 m (3.28 ft)
TPE 374, 2 m (6.6 ft)
TPE 374, 2 m (78.7 in)
Dimensione della confezione:
pezzi 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipi di connettore Dimensioni dei tubi unitSize
17850577 Luer to MPC TPE 374, 1 m (3.28 ft) pezzi 10
17870647 Luer to MPC TPE 374, 2 m (6.6 ft) pezzi 10
17830577 MPX to MPC TPE 374, 2 m (78.7 in) pezzi 10
17870577 MPX to MPC TPE 374, 2 m (6.6 ft) pezzi 10
17860577 MPX to MPC C-Flex 374, 2 m (78.7 in) pezzi 10
17890647 MPX to MPC C-Flex 374, 2 m (6.6 ft) pezzi 10
17800587 MPX/MPC TPE 374, 2 m (78.7 in) pezzi 10
17800657 MPX/MPC TPE 374, 2 m (6.6 ft) pezzi 10
17830657 MPC/MPX TPE 374, 2 m (78.7 in) pezzi 10
17840657 MPC/MPX TPE 374, 2 m (6.6 ft) pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Codice prodotto. 17850577 Fornitore Thermo Scientific™ N. del fornitore SH31121.01
 Articolo soggetto a restrizioni
Spedizione stimata: 12-11-2026
Aggiungi al carrello
Aggiungi al carrello
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
TRUSTED_SUSTAINABILITY
Specifica
Dimensions 1/8 in. ID x 1/4 in. OD to 3/8 in. ID x 5/8 in. OD
Stile dei tubi 3.22 mm (1/8 in.) ID x 6.35 mm (1/4 in.) OD to 9.52 mm (3/8 in.) ID x 15.88 mm (5/8 in.) OD; TPE 374; 1 m (3.28 ft); Luer to MPC
Tipi di connettore Luer to MPC
Product Type Adapter Set
Dimensioni dei tubi TPE 374, 1 m (3.28 ft)