Learn More
Thermo Scientific™ Fluid Transfer Assembly, Y Set
Codice prodotto. 17890627 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Stile dei tubi:
3 x TPE 374
Tipi di connettore:
Luer
MPX body-insert
MPX insert-body
Dimensions:
1/2 in. ID x 3/4 in. wall
1/4 in. ID x 7/16 in. wall
1/8 in. ID x 1/4 in. wall
3/8 in. ID x 5/8 in. wall
Dimensione della confezione:
pezzi 10
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Stile dei tubi Tipi di connettore Dimensions Dimensione della confezione
17890627 3 x TPE 374 MPX body-insert 3/8 in. ID x 5/8 in. wall pezzi 10
17800637 3 x TPE 374 MPX body-insert 1/4 in. ID x 7/16 in. wall pezzi 10
17830687 3 x TPE 374 MPX insert-body 1/4 in. ID x 7/16 in. wall pezzi 10
17810637 3 x TPE 374 MPX insert-body 3/8 in. ID x 5/8 in. wall pezzi 10
17870627 3 x TPE 374 MPX body-insert 1/2 in. ID x 3/4 in. wall pezzi 10
17820637 3 x TPE 374 Luer 1/8 in. ID x 1/4 in. wall pezzi 10
17840687 3 x TPE 374 MPX insert-body 1/2 in. ID x 3/4 in. wall pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Codice prodotto. 17890627 Fornitore Thermo Scientific™ N. del fornitore SH31106.01
  • ONLINE EXCLUSIVE PRICE
    625.00€
    511.76€ / pezzi 10
    Risparmia 113.24€
    18% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 03-03-2027
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
TRUSTED_SUSTAINABILITY
Specifica
Dimensions 3/8 in. ID x 5/8 in. wall
Stile dei tubi 3 x TPE 374
Tipi di connettore MPX body-insert
Product Type Y Set
Dimensioni dei tubi 2 x 20 in. to 20 in. (Length)