Learn More
Flumioxazin (free amine) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30309306
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309306 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30309306 Supplier Dr. Ehrenstorfer Supplier No. DREA13725200AL100
 Restricted Item
Spedizione stimata: 08-01-2027
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Flumioxazin (free amine)
CAS 103361-42-8
Analyte or Component Names Flumioxazin (free amine)
Shipping Condition Room Temperature
Molecular Formula C11 H9 F N2 O2
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym 2H-1,4-Benzoxazin-3(4H)-one, 6-amino-7-fluoro-4-(2-propyn-1-yl)-, 2H-1,4-Benzoxazin-3(4H)-one, 6-amino-7-fluoro-4-(2-propynyl)- (9CI), 6-Amino-7-fluoro-4-(2-propyn-1-yl)-2H-1,4-benzoxazin-3(4H)-one
InChI Formula InChI=1S/C11H9FN2O2/c1-2-3-14-9-5-8(13)7(12)4-10(9)16-6-11(14)15/h1,4-5H,3,6,13H2
SMILES Nc1cc2N(CC#C)C(=O)COc2cc1F
IUPAC Name 6-amino-7-fluoro-4-prop-2-ynyl-1,4-benzoxazin-3-one
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 220.2
Quantity 1 mL
Formula Weight 220.065 g/mol