Learn More
Abcam Fluo-5 AM, Fluorescent labeling reagent (Fluo-4 analogue)
Codice prodotto. 32119138 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
10 x 50 μg
Dimensione della confezione:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32119138 10 x 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32119138 Fornitore Abcam N. del fornitore ab24108310x50ug
 Articolo soggetto a restrizioni
Spedizione stimata: 02-12-2026
MW 740.6 Da, Purity >98%. Fluo-5 AM is an analogue of Fluo-4 with reduced calcium binding affinity (Kd = 2.3 μM). Suitable for detecting intracellular calcium levels in the range 1 μM to 1 mM. Cells can be loaded with Fluo-5 AM by adding the dissolved indicator directly to cells in cul...
MW 740.6 Da, Purity >98%. Fluo-5 AM is an analogue of Fluo-4 with reduced calcium binding affinity (Kd = 2.3 μM). Suitable for detecting intracellular calcium levels in the range 1 μM to 1 mM. Cells can be loaded with Fluo-5 AM by adding the dissolved indicator directly to cells in culture dishes.
Fluo-5 AM has excitation and emission wavelengths at 494 and 516 nm respectively. Compatible with excitation by argon-ion lasers at 488 nm. Fluorescence inceases by >100-fold upon binding calcium.
.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 10 x 50 μg
Status giuridico RUO