Learn More
Mettler Toledo™ Fluoride Membrane Kit
Product Code. 10107211
Change view
Click to view available options
Material:
Solid State Membrane
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10107211 Solid State Membrane 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10107211 Supplier Mettler Toledo™ Supplier No. 51340008
  • ONLINE EXCLUSIVE PRICE
    720.00€
    611.69€ / 1 sets
    Risparmia 108.31€
    15% Sconto
View all promotions
In stock
Add to basket
Edge
Specifications
For Use With (Application) In gold Fluoride ISE
Material Solid State Membrane
Temperature (Metric) Operating 0°C to 80°C
pH Range 4 to 10