Learn More
Hanna Instruments™ Fluoride Reagent Kit
Product Code. 11703353
Change view
Click to view available options
Product Type:
Fluoride Reagent Kit
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11703353 Fluoride Reagent Kit Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11703353 Supplier Hanna Instruments™ Supplier No. HI9373901
  • ONLINE EXCLUSIVE PRICE
    364.55€
    185.40€ / pezzi 100
    Risparmia 179.15€
    49% Sconto
 Restricted Item
Spedizione stimata: 28-12-2026
Specifications
Product Type Fluoride Reagent Kit
compliance-icons compliance-icons
Product Identifier
  • X100 FLUORIDE REAGENT FOR 16426
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid