Learn More
Cole-Parmer™ FMI Series RH Valveless Piston Pump Head
Codice prodotto. 15592173 Sfoglia Tutto Masterflex Prodotti
Cambia vista
Click to view available options
Portata:
10 to 100μL
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Portata Dimensione della confezione
15592173 10 to 100μL Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15592173 Fornitore Cole-Parmer™ N. del fornitore 0711765
  • ONLINE EXCLUSIVE PRICE
    1865.00€
    1298.44€ / pezzi 1
    Risparmia 566.56€
    30% Sconto
Spedizione stimata: 04-01-2027
Valveless piston pumps are low maintenance and suffer fewer piston seizures or clogs. With a single moving part, there is less drift over time, offering millions of cycles of reliable, repeatable pumping.
  • Small displacement provides excellent precision
  • Fully adjustable from zero to maximum
Specifica
Descrizione Valveless Piston Pump Head
Portata 10 to 100μL
Pressione massima 100 psi