Learn More
Uvex™ FR Shirt 7421-Dark Gray
Catalog Number 17276691
Click to view available options
Size:
37/38
39/40
41/42
43/44
45/46
47/48
49/50
This shirt is the perfect supplement to various jackets and trousers from the uvex range of protective clothing. Best-possible protection - even under your jacket.
  • Material 55% modacryl FR/ 49 % cotton, 2 % antistatic fibres, 165 g/m²
  • Well protected: men's shirt with antistatic protection for heatexposed industrial workers
  • Turn down collar with neckband
  • Visible button strip
  • 1 breast pocket with flap and button
  • Long arm, cuff with adjustable button closure
  • Certified according to EN ISO 11612 A1, B1, C1, F1 EN 1149-5:2008 IEC 61482-1-2 test category 1 sizes 37/38 - 49/50
  • Care instructions: 60°C washable, no chlorinate, dry-cleaning: P, iron temperature: II-points tumbler: I-point
Size 45/46
Sesso Men's
Colore Gray
Tipo di chiusura Press studs
Monouso No