Learn More
Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules, BreakApart frame
Descrizione
Use Thermo Scientific™ Frames and Accessories for Immuno Breakable Modules with Nunc™ C8 Breakable Modules. These frames include tools to facilitate removing the strips.
Warranty: 90 days
Specifica
Specifica
| Formato | Strip Plate |
| Descrizione | High optical quality for reliable results |
| Matrice | 8 x 12 |
| Imballaggio | 60 Cases, 5 Pk. |
| Colore | White |
| Product Type | Frame |
| Materiale | Polystyrene |
| No. of Wells | 96 |
| Forma pozzetto | Curved Edges, Flat Bottom (C-well) |
| Da utilizzare con (applicazione) | For solid phase immuno assays |
| Vedi altri risultati |