Learn More
Eppendorf™ Freezer Rack: CryoCube™ FC660 series, MTP, side access, stainless steel
Descrizione
Freezer Rack: CryoCube™ FC660 series, MTP, side access, stainless steel- Rack for MTP (18 mm)- Max. MTP size [mm]: 86 x 128- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 133 x 91 x 746 (5.2 x 3.6 x 29.4 in)- Max. MTP number per rack: 37 (37 x 1)- Stainless steel- For ULT freezer: CryoCube™ FC660/ FC660h (max. 60 racks)- For ULT freezer: CryoCube™ FC660/ FC660h (max. 2,220 MTP)
Specifica
Specifica
| Materiale | Stainless Steel |
| Product Type | Freezer Rack |