Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, 2 in/53 mm
Descrizione
Freezer Rack: Innova™ C580, C760, locking rod, 2 in/53 mm, side access, stainless steel- Rack for 53 mm/2 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 148 x 141 x 776 (5.8 x 5.6 x 30.6)- Max. box number per rack: 14 (14 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks)- For ULT freezer: Innova C580 (max. 448 boxes), Innova C760 (max. 560 boxes)- With locking rod for increased sample safety
Specifica
Specifica
| Product Type | Freezer Rack |