Learn More
Eppendorf™ Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, 3 in/76 mm
Descrizione
Freezer Rack: Innova™ C580, C760, CryoCube™ FC660 series, 3 in/76 mm, side access, stainless steel- Rack for 76 mm/3.0 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 141 x 141 x 726 (5.6 x 5.6 x 28.6 in)- Max. box number per rack: 9 (9 x 1)- Stainless steel- For ULT freezer: Innova C580 (max. 32 racks), Innova C760 (max. 40 racks), CryoCube™ FC660 series (max. 40 racks)- For ULT freezer: Innova C580 (max. 288 boxes), Innova C760 (max. 360 boxes), CryoCube™ FC660 series (max. 360 boxes)
Specifica
Specifica
| Product Type | Freezer Rack |