Learn More
Eppendorf™ Freezer Rack: Innova™ U101, CryoCube™ F101h, 2.5 in/64 mm
Descrizione
Freezer Rack: Innova™ U101, CryoCube™ F101h, 2.5 in/64 mm, side access, stainless steel- Rack for 64 mm/2.5 in boxes (5 mL tubes)- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 290 x 142 x 264 (11.4 x 5.6 x 10.4 in)- Max. box number per rack: 8 (4 x 2)- Stainless steel- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 6 racks)- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 48 boxes)
Specifica
Specifica
| Product Type | Freezer Rack |