Learn More
Eppendorf™ Freezer Rack: Innova™ U101, CryoCube™ F101h, 2 in/53 mm
Descrizione
Freezer Rack: Innova™ U101, CryoCube™ F101h, 2 in/53 mm, side access, stainless steel- Rack for 53 mm/2 in boxes- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 290 x 142 x 279 (11.4 x 5.6 x 11 in)- Max. box number per rack: 10 (5 x 2)- Stainless steel- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 6 racks)- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 60 boxes)
Specifica
Specifica
| Product Type | Freezer Rack |