Learn More
Eppendorf™ Freezer Rack: Innova™ U101, CryoCube™ F101h, 4 in/102 mm
Descrizione
Freezer Rack: Innova™ U101, CryoCube™ F101h, 4 in/102 mm, drawer, stainless steel- Rack for 102 mm/4 in boxes- Max. box size [mm]: 136 x 136- Drawer rack- Ext. dimensions of rack (DxWxH) [mm]: 308 x 145 x 216 (12.1 x 5.7 x 8.5 in)- Max. box number per rack: 4 (2 x 2)- Stainless steel- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 6 racks)- For ULT freezer: CryoCube™ F101h/ Innova™ U101 (max. 24 boxes)
Specifica
Specifica
| Product Type | Freezer Rack |