Learn More
Eppendorf™ Freezer Rack: Innova™ U360, U535, U725/-G, CryoCube™ F740 series (3-comp., Std),
Product Code. 15753235
Product Code. 15753235 Supplier Eppendorf™ Supplier No. 6001011910
 Restricted Item
In stock
Comprehensive storage of samples in a ULT freezer is based on storage boxes. The boxes should be stored in dedicated freezer racks. These metal racks enable optimal use of space as well as easy relocation of sample boxes.
Freezer Rack: Innova™ U360, U535, U725, U725-G, CryoCube™ F740 series (3-compartment, Standard), 2.5 in/64 mm, side access, stainless steel- Rack for 64 mm/2.5 in boxes (5 mL tubes)- Max. box size [mm]: 136 x 136- Side access rack- Ext. dimensions of rack (DxWxH) [mm]: 569 x 139 x 394 (22.4 x 5.5 x 15.5 in)- Max. box number per rack: 24 (4 x 6)- Stainless steel- For ULT freezer: Innova U360 (max. 9 racks), Innova U535 (max. 12 racks), Innova U725 and U725-G (max. 18 racks), CryoCube F740 series (3-compartment version; max. 18 racks)- For ULT freezer: Innova U360 (max. 216 boxes), Innova U535 (max. 288 boxes), Innova U725 and U725-G (max. 432 boxes), CryoCube F740 series (3-compartment version; max. 432 boxes)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Freezer Rack