Learn More
Labconco™ FreeZone™ 4.5 L -105°C Benchtop Freeze Dryer, 230 V Model
BIOPHARMA
Codice prodotto. 15772847 Sfoglia Tutto Labconco Corporation Prodotti
Cambia vista
Click to view available options
Tipo di spina:
Schuko
Include:
Non-coated Stainless Steel Collector
PTFE-coated Stainless Steel Collector
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo di spina Include Dimensione della confezione
15772847 Schuko PTFE-coated Stainless Steel Collector Pezzo
15752847 Schuko Non-coated Stainless Steel Collector Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15772847 Fornitore Labconco™ N. del fornitore 720402030
  • ONLINE EXCLUSIVE PRICE
    20930.00€
    16233.60€ / pezzi 1
    Risparmia 4696.40€
    22% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
Spedizione stimata: 04-01-2027
Includes
PTFE-coated collector
Requires
Drying accessory, vacuum pump, glassware, adapters
FreeZone 4.5 Liter Benchtop Freeze Dryers are designed for lyophilizing light to moderate sample loads. The compact design is ideal for labs with limited floor space. New Lyo-Works™ OS provides flexibility and convenience in an easy-to-use, touch screen display.
  • Upright stainless steel collector coil holds 4.5 liters of ice before defrosting
  • -105° C (-157° F) at 60 Hz or -102° C (-152° F) at 50 Hz collector is suitable for low eutectic point samples including acetonitrile and dilute methanol and ethanol
  • Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection
  • Environmentally-friendly HCFC/CFC-free refrigeration system
  • Internal coil maximizes vapor-trapping efficiency
  • Optional PTFE-coated collectors withstand corrosive compounds
  • Lyo-Works OS provides a real time display of collector temperature and vacuum level
  • 5 in. full color touch screen display
  • Flexible programming and control of optional drying accessories
  • Optional End-Zone™ end point detection for flask freeze drying
  • One-touch data logging and display provides information to maximize sample quality and minimize run time
  • Customizable settings include language, time zone, temperature and vacuum
  • Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses
  • FreeZone exclusive features protect your vacuum pump from costly damage: A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run
Vacuum pump with a displacement of at least 98 liters/minute, drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).
Compliance
All models conform to UL Standard 61010-1 (60 Hz models), CAN/CSA C22.2 No. 61010-1 (60 Hz models), and CE Conformity marking (230 V, 50 Hz models).
Warranty
Un anno
Specifica
Tipo Freeze Dryer/Lyophilizer
Capacità (sistema metrico) 4.5 L (before defrosting)
N. di ripiani 0
Temperature Range (Metric) -105°C
Intervallo di temperatura (sistema imperiale) -157°F
Materiale Steel
Certificazione/Conformità UL, CAN/CSA, CE
Include PTFE-coated Stainless Steel Collector
Profondità (sistema imperiale) 26.5 in.
Profondità (sistema metrico) 67.3 cm
Larghezza (sistema imperiale) 22.5 in.
Larghezza (sistema metrico) 57.1 cm
Altezza (sistema imperiale) 18.5 in.
Altezza (sistema metrico) 46.5 cm
Amperaggio 10 A
Tipo di spina Schuko
Tensione 230 V
Frequenza 60 Hz
Peso di spedizione (sistema imperiale) 172 lb.
Peso di spedizione (sistema metrico) 78 kg