Learn More
Labconco™ Liofilizzatore da banco FreeZone™ 4.5 L -50 °C, modelli 230V
BIOPHARMA
Codice prodotto. 15772727 Sfoglia Tutto Labconco Corporation Prodotti
Cambia vista
Click to view available options
Tipo di spina:
Schuko
Include:
Non-coated Stainless Steel Collector
PTFE-coated Stainless Steel Collector
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo di spina Include unitSize
15772727 Schuko Non-coated Stainless Steel Collector Pezzo
15782727 Schuko PTFE-coated Stainless Steel Collector Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15772727 Fornitore Labconco™ N. del fornitore 700401030
Please to purchase this item. Need a web account? Register with us today!
Includes
Stainless steel collector
Requires
Drying accessory, vacuum pump, glassware, adapters
I crioessiccatori da banco FreeZone da 4.5 litri sono progettati per la liofilizzazione di carichi di campioni sia leggeri che moderati. Il design compatto è ideale per i laboratori con spazio al suolo limitato. Il nuovo Lyo-Works™ OS offre flessibilità e comodità in un display con schermo touchscreen di facile utilizzo.
  • Upright stainless steel collector coil holds 4.5 liters of ice before defrosting
  • -50° C collector is suitable for aqueous samples
  • Features include hot gas defrost, side-mounted USB port and rear-mounted Ethernet connection
  • Environmentally-friendly HCFC/CFC-free refrigeration system
  • Internal coil maximizes vapor-trapping efficiency
  • Optional PTFE-coated collectors withstand corrosive compounds
  • Lyo-Works OS provides a real time display of collector temperature and vacuum level
  • 5 in. full color touch screen display
  • Flexible programming and control of optional drying accessories
  • Optional End-Zone™ end point detection for flask freeze drying
  • One-touch data logging and display provides information to maximize sample quality and minimize run time
  • Customizable settings include language, time zone, temperature and vacuum
  • Alerts and feedback of current operating parameters can be sent automatically to user-entered email addresses
  • FreeZone exclusive features protect your vacuum pump from costly damage: A vacuum break valve bleeds air into the system when power to the freeze dryer or vacuum pump is shut off for more than 5 minutes. A moisture sensor prevents refrigeration or vacuum start-up when moisture is detected in the drain line. Vacuum pump is automatically shut down when a leak is present in initial pull down or during run
Vacuum pump with a displacement of at least 98 liters/minute, drying accessory and freeze dry glassware (if not bulk freeze drying) are required (not included).
Compliance
Tutti i modelli sono conformi allo standard UL 61010-1 (modelli da 60 Hz), CAN/CSA C22.2 n. 61010-1 (modelli da 60 Hz) e riportano il contrassegno di conformità CE (modelli da 230 V, 50 Hz).
Warranty
Un anno
Specifica
Tipo Freeze Dryer/Lyophilizer
Capacità (sistema metrico) 2.5 L (before defrosting)
N. di ripiani 0
Temperature Range (Metric) -50°C
Intervallo di temperatura (sistema imperiale) -58°F
Materiale Steel
Certificazione/Conformità UL, CAN/CSA, CE
Include Non-coated Stainless Steel Collector
Profondità (sistema imperiale) 22.4 in.
Profondità (sistema metrico) 56.8 cm
Larghezza (sistema imperiale) 16.5 in.
Larghezza (sistema metrico) 41.9 cm
Altezza (sistema imperiale) 18.9 in.
Altezza (sistema metrico) 48 cm
Amperaggio 7 A
Tipo di spina Schuko
Tensione 230 V
Frequenza 60 Hz
Peso di spedizione (sistema metrico) 49.4 kg