Learn More
Labconco™ FreeZone™ Stoppering Tray Dryers, Isolation Valve, 230V Models
Codice prodotto. 15762867 Sfoglia Tutto Labconco Corporation Prodotti
Cambia vista
Click to view available options
Tipo di spina:
Schuko
Include:
3 Shelves, Isolation Valve
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo di spina Include Dimensione della confezione
15762867 Schuko 3 Shelves, Isolation Valve Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15762867 Fornitore Labconco™ N. del fornitore 794801230
  • ONLINE EXCLUSIVE PRICE
    38450.00€
    29826.12€ / pezzi 1
    Risparmia 8623.88€
    22% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 29-12-2026
Includes
3 shelves, Isolation Valve
Requires
Freeze dryer, vacuum pump
Provides precise control and program flexibility for consistent performance. Labconco™ FreeZone™ Stoppering Tray Dryers, Isolation Valve, 230V Models are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. They offer options for built-in 6-port manifold and isolation valve.
  • Three 16.5 x 12 in. temperature-controlled (±1° C), adjustable shelves with sample temperature sensors
  • Chamber pre-freezes samples to save time and money by eliminating need for separate freezer and product transfer
  • Up to 594 sq. inches of shelf space
  • Circulating fluid in shelves ranges from −40 and +40° C
  • Environmentally-friendly HCFC/CFC-free refrigeration system
  • Built-in pneumatic mechanism stoppers containers on all three shelves
  • Clear acrylic viewing door provides complete visibility of shelves
  • 1/8 in. OD backfill port introduces sterile air or inert gas from outside source to chamber, protecting samples from atmospheric moisture and contaminants
  • Lyo-Works™ OS provides real time display of freeze dryer and tray dryer parameters on 5 in. full color, capacitive touch screen on connected FreeZone 6, 12, or 18L Freeze Dryer (sold separately)
  • Data may be exported via USB or Ethernet
  • Microprocessor-based controller allows up to 30 different temperature programs to be stored and repeated, each using up to 16 different steps
  • Display provides navigation to manual or program mode, probe temperatures, vacuum in mBar, Torr or Pa, step of program in progress, time remaining in present step, and end of program, as well as alarms
  • Alerts and current operating parameter reading can be sent automatically to user-entered email addresses
  • Optional 6-Port Tray Dryer Manifold accommodates either 1/2 in. or 3/4 in. flask adapters and connect to flasks and other glassware
  • Optional built-in isolation valve separates Stoppering Tray Dryer from freeze dryer to shorten time necessary to reestablish working vacuum levels and to provide means for determining end point
  • Freeze dryer, vacuum pump with displacement of at least 163L/minute, and freeze dry glassware (if not bulk freeze drying) are required (not included)
Compliance
All models conform to UL Standard 61010-1 (60 Hz models), CAN/CSA C22.2 No. 61010-1 (60 Hz models), and CE Conformity marking (230V, 50 Hz models).
Warranty
Un anno
Specifica
Tipo Stoppering Tray Dryer
N. di ripiani 3
Temperature Range (Metric) -40°C to +40°C
Materiale Steel
Certificazione/Conformità UL, CAN/CSA, CE
Include 3 Shelves, Isolation Valve
Profondità (sistema imperiale) 24.6 in.
Profondità (sistema metrico) 62.4 cm
Larghezza (sistema imperiale) 32.4 in.
Larghezza (sistema metrico) 82.2 cm
Altezza (sistema imperiale) 27.1 in.
Altezza (sistema metrico) 68.8 cm
Amperaggio 9A
Tipo di spina Schuko
Tensione 230 V
Frequenza 60 Hz
Peso di spedizione (sistema imperiale) 431 lb.
Peso di spedizione (sistema metrico) 195.49 kg