missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Fyn Overexpression Lysate

Codice prodotto. 18274384 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mg
Dimensione della confezione:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18274384 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18274384 Fornitore Novus Biologicals™ N. del fornitore NBL110885
 Articolo soggetto a restrizioni
Spedizione stimata: 19-10-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
The Fyn Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifica

Linea cellulare The lysate was created in HEK293T cells, using plasmid ID RC224691 and based on accession number NM_237. The protein contains a C-terminal DDK tag.
Da utilizzare con (applicazione) Western Blot
Quantità 0.1 mg
Tampone di conservazione RIPA buffer
Purezza Protein
Test di specificità Homo sapiens FYN oncogene related to SRC, FGR, YES (FYN), transcript variant 1, mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.