missing translation for 'onlineSavingsMsg'
Learn More

GALE Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA552423

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15780371

  • 514.00€ / 100µL
Spedizione stimata: 10-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: QGIPNNLMPY VSQVAIGRRE ALNVFGNDYD TEDGTGVRDY IHVVDLAKGH IAALRKLKEQ CGCRIYNLGT GTGYSVLQMV QAMEKASGKK IPYKVVARRE GDVAA Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

GALE Polyclonal antibody specifically detects GALE in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifica

Specifica

GALE
Polyclonal
Unconjugated
GALE
2310002A12Rik; AI323962; EGK_00373; galactose-4-epimerase, UDP; galactose-4-epimerase, UDP-; galactowaldenase; Gale; SDR1E1; short chain dehydrogenase/reductase family 1E, member 1; UDP galactose-4'-epimerase; UDP-galactosamine 4-epimerase; UDP-galactose 4-epimerase; UDP-galactose-4-epimerase; UDP-GalNAc 4-epimerase; UDP-GlcNAc 4-epimerase; UDP-glucose 4-epimerase; UDP-N-acetylgalactosamine 4-epimerase; UDP-N-acetylglucosamine 4-epimerase
Rabbit
Antigen affinity chromatography
RUO
114860, 2582, 74246
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P18645, Q14376, Q8R059
GALE
Recombinant protein corresponding to Human GALE
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only