Learn More
Fisherbrand™ Gamma Base Plates
Product Code. 12347197 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Height (Metric):
410 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
12347197 410 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12347197
Spedizione stimata: 05-04-2027
These rectangular base plates provide bench protection and can be used in conjunction with any shields. The plate is manufactured with cushioned anti-slip feet.
  • Rectangular can be used with any shields and has cushioned anti slip feet 450mm (w) x 410mm (h) shield
  • These rectangular base plates provide bench protection and can be used in conjunction with any shields
  • The plate is manufactured with cushioned anti-slip feet Gamma Base Plates
  • Available in standard 10mm beta-protecting acrylic, 12mm gamma-attenuating lead acrylic
  • Cleaver Scientific's comprehensive range of Radiation Safety Products comprises a large selection of shields,
  • boxes, waste bins, trays, plus assorted accessories and cabinets
Specifications
Product Type Gamma Base Plate
Width (Metric) 450 mm
Height (Metric) 410 mm