Learn More
gamma-Linolenic Acid Ethyl Ester, TRC
Specifica
Specifica
| Materiale o nome chimico | gamma-Linolenic Acid Ethyl Ester |
| CAS | 31450-14-3 |
| Formula molecolare | C20H34O2 |
| Quantità | 1 g |
| Sinonimo | (6Z,9Z,12Z)-6,9,12-Octadecatrienoic Acid Ethyl Ester, (Z,Z,Z)-6,9,12-Octadecatrienoic Acid Ethyl Ester, Ethyl γ-Linolenate, Ethyl γ-Linolenic Acid |
| InChI Formula | InChI=1S/C20H34O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h8-9,11-12,14-15H,3-7,10,13,16-19H2,1-2H3/b9-8-,12-11-,15-14- |
| SMILES | O=C(OCC)CCCC/C=C\C/C=C\C/C=C\CCCCC |
| IUPAC Name | ethyl (6Z,9Z,12Z)-octadeca-6,9,12-trienoate |
| Molecular Weight (g/mol) | 306.48 |
| Peso formulazione | 306.2559 |