missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
GAS2 Polyclonal antibody specifically detects GAS2 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifica
Specifica
| Antigene | GAS2 |
| Applicazioni | Western Blot, Immunoprecipitation |
| Classificazione | Polyclonal |
| Coniugato | Unconjugated |
| Diluizione | Western Blot 1:500-1:1000, Immunoprecipitation 1-2 μL/mg of lysate |
| Formulazione | 0.2 um filtered solution in PBS |
| Alias gene | GAS-2, growth arrest-specific 2, growth arrest-specific protein 2, MGC32610 |
| Specie ospite | Rabbit |
| Immunogeno | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human GAS2. |
| Metodo di purificazione | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?