missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
GATA-3 Monoclonal specifically detects GATA-3 in Human samples. It is validated for ELISA∣Flow Cytometry∣Western Blot.
Specifica
Specifica
| Antigene | GATA-3 |
| Applicazioni | ELISA, Flow Cytometry, Western Blot |
| Classificazione | Monoclonal |
| Clone | GATA3/2446 |
| Coniugato | PE |
| Formulazione | PBS |
| Alias gene | 131320, GATA binding protein 3, GATA-binding factor 3, GATA-binding protein 3, HDRMGC5199, MGC2346, MGC5445, trans-acting T-cell-specific transcription factor GATA-3 |
| Specie ospite | Mouse |
| Immunogeno | Recombinant fragment of human GATA-3 protein (around aa 357-436) (exact sequence is proprietary) (Uniprot: P23771) |
| Metodo di purificazione | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?