missing translation for 'onlineSavingsMsg'
Learn More

GBL Antibody, mFluor Violet 450 SE, Novus Biologicals™

Codice prodotto. 18525253 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.1 mL
Dimensione della confezione:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18525253 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18525253 Fornitore Novus Biologicals N. del fornitore NBP298484MFV450

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

GBL Polyclonal antibody specifically detects GBL in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifica

Antigene GBL
Applicazioni Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classificazione Polyclonal
Coniugato mFluor Violet 450 SE
Diluizione Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulazione 50mM Sodium Borate
Alias gene G protein beta subunit-like, Gable, GbetaL, GBLPop3, Lst8, LST8MGC111011, Mammalian lethal with SEC13 protein 8, mLST8, MTOR associated protein, LST8 homolog (S. cerevisiae), POP3, Protein GbetaL, target of rapamycin complex subunit LST8, TORC subunit LST8, WAT1
Specie ospite Rabbit
Immunogeno Produced in rabbits immunized with E. coli-derived Human GBL fragment.
Metodo di purificazione Antigen and protein A Affinity-purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Cancer, Hypoxia, mTOR Pathway
Primario o secondario Primary
ID gene (immissione) 64223
Target Species Human
Content And Storage Store at 4C in the dark.
Forma Purified
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.