Learn More
Trajan Scientific and Medical™ GC Inlet Liner Large Volume Injection (Lvi) Sintered Glass For Agilent Gerstel Cis 4 Injector 1.8mm I.D.
Descrizione
- Each batch of liners tested for activity
- Manufactured from sintered glass
- Color coded by geometry for easy identification
- For use with Agilent Technologies instruments
Specifica
Specifica
| Product Type | Large Volume Injection (LVI) Inlet Liner |
| Descrizione | Sintered Glass |
| Quantità | 5/Pk. |
| Lunghezza (sistema metrico) | 71 mm |
| Diametro (sistema metrico) interno | 1.8 mm |
| Diametro (sistema metrico) esterno | 3 mm |
| Da utilizzare con (apparecchiatura) | Gerstel CIS 4 Injector |