Learn More
Thermo Scientific™ GC Installation Kit
Product Code. 10589165 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Gas Chromatography Systems
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10589165 Gas Chromatography Systems Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10589165 Supplier Thermo Scientific™ Supplier No. 60180888
Spedizione stimata: 01-12-2026
Add a GC system to your lab bench with the Thermo Scientific™ GC Installation Kit. The required tubing and fittings are conveniently supplied in this kit.
Add a GC system to your lab bench with the Thermo Scientific™ GC Installation Kit. The required tubing and fittings are conveniently supplied in this kit.
Includes:
Tubing cutter; 1/8 x 1/4in. reamer; 7/16in. wrench; 1/2in. wrench; 1/8in. brass tees, 4; 1/8in. brass nuts, 10; Brass front and back ferrules, 10; 15.2m instrument-grade, cleaned 1/8in. copper tubing
TRUSTED_SUSTAINABILITY
Specifications
Type GC Installation Kit
Description GC Installation Kit
For Use With (Application) GC
Quantity Each
For Use With (Equipment) Gas Chromatography Systems