Learn More
Biochrom™ Datrys Software
Datrys software enables PC control of a range of GE spectrophotometers
Marca:  Biochrom™ 29003624
Dettagli aggiuntivi : Peso : 0.05000kg
Codice prodotto. 12805843
  • 2028.00€ / Pezzo
Spedizione stimata
22-02-2021
Aggiungi al carrello
Descrizione
Descrizione
  • Four software packages are available, depending on your user requirements, and it is simple to upgrade should your requirements change
Choose a package that suits your needs:
  • Datrys Lite: for quick read and fast scanning only
  • Datrys Standard: for all routine measurements
  • Datrys Life Science: for nucleic acids, proteins, and cell density measurements
  • Datrys CFR: for labs requiring 21 CFR part 11 compatibility
  • Offering familiar look and feel of Microsoft™ Office 2007, and is compatible with Windows™ XP, Windows Vista™ and Windows™ 7 operating systems
  • Data export options include Microsoft Word™ and Excel plus Adobe™ PDF formats
  • Datrys comes as standard on Ultrospec 7000PC, Ultrospec 8000 PC, Ultrospec 8000 stand-alone and Ultropsec 9000PC and stand-alone versions
  • Compatible with NanoVue , NanoVue Plus, GeneQuant 100 , GeneQuant 1300, Ultrospec 2100, Ultrospec 3100, Ultrospec 3300, Ultrospec 4300, Ultrospec 5300 and Ultrospec 6300
Specifica
Specifica
For labs requiring 21 CFR part 11 compatibility
A range of GE spectrophotometers
Datrys CFR Software