Learn More
Biochrom™ Datrys Software
Datrys software enables PC control of a range of GE spectrophotometers
Marca: Biochrom™ 29003624
Dettagli aggiuntivi : Peso : 0.05000kg
Descrizione
- Four software packages are available, depending on your user requirements, and it is simple to upgrade should your requirements change
- Datrys Lite: for quick read and fast scanning only
- Datrys Standard: for all routine measurements
- Datrys Life Science: for nucleic acids, proteins, and cell density measurements
- Datrys CFR: for labs requiring 21 CFR part 11 compatibility
- Offering familiar look and feel of Microsoft™ Office 2007, and is compatible with Windows™ XP, Windows Vista™ and Windows™ 7 operating systems
- Data export options include Microsoft Word™ and Excel plus Adobe™ PDF formats
- Datrys comes as standard on Ultrospec 7000PC, Ultrospec 8000 PC, Ultrospec 8000 stand-alone and Ultropsec 9000PC and stand-alone versions
- Compatible with NanoVue , NanoVue Plus, GeneQuant 100 , GeneQuant 1300, Ultrospec 2100, Ultrospec 3100, Ultrospec 3300, Ultrospec 4300, Ultrospec 5300 and Ultrospec 6300
Specifica
For labs requiring 21 CFR part 11 compatibility | |
A range of GE spectrophotometers | |
Datrys CFR Software |