Learn More
Thermo Scientific™ Scala DNA a intervallo alto GeneRuler™
Codice prodotto. 10374880 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
5 x 50 μg
50 µg
Dimensione della confezione:
50µg
pezzi 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
10374880 50 µg 50µg
11843993 5 x 50 μg pezzi 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 10374880 Fornitore Thermo Scientific™ N. del fornitore SM1351
Follow the Workflow - Explore the Savings
Usa il codice promozionale 27967
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Edge
Aggiungi al carrello
Perform fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels.
GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels (in 1.5hour, 0.4%agarose gel). The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler High Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol in the Resources.)
The ladder is supplied with 6X DNA Loading Dye for sample DNA.
GeneRuler High Range DNA Ladder is recommended for fast sizing and approximate quantification of high molecular weight double-stranded DNA on agarose gels (in 1.5 hours, 0.4% agarose gel). The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler High Range DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase.The ladder is supplied with 6X DNA Loading Dye for sample DNA.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Supplied with loading dye for sample DNA
  • Ideal for fast sizing of high molecular weight DNA fragments
TRUSTED_SUSTAINABILITY
Specifica
Concentrazione 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Ready to Load No
Intervallo dimensioni 10171 to 48502 bp
Compatibilità gel Agarose Gels
Quantità 50 µg
Product Line GeneRuler
Product Type High Range DNA Ladder