Learn More
Cole Parmer GFS Chemicals Turbidity Primary Standard Solution
Product Code. 15285662 Shop All Cole-Parmer Products
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15285662 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15285662 Supplier Cole Parmer Supplier No. 0839109
  • ONLINE EXCLUSIVE PRICE
    308.00€
    299.30€ / pezzi 1
    Risparmia 8.70€
    2% Sconto
 Restricted Item
Spedizione stimata: 12-01-2027
GFS Chemicals Turbidity Primary Standard Solution can be used to calibrate nephelometers or turbidimeters.
  • EPA-approved polymeric standards outlast Formazin standards.
  • Suspended polymeric particles virtually never settle or clump.
  • Solutions maintain NTU values within ±1% for one year.
  • NTU Value: 0.5
Specifications
Type Turbidity Meter Standards and Calibrators
Volume (English) 33.8 oz.
Volume (Metric) 1 L