Learn More
3M™ GH4 Chinstrap
Product Code. 15729967 Shop All 3M Products
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15729967 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15729967 Supplier 3M™ Supplier No. 7000108175
  • ONLINE EXCLUSIVE PRICE
    92.00€
    74.43€ / pezzi 10
    Risparmia 17.57€
    19% Sconto
Spedizione stimata: 05-02-2027
3M™ GH4 Chinstrap is a replacement part for 3M™ Hard Hat Series G3000.
  • Replacement 3-point Chinstrap
  • Suitable for use with 3M Hard Hat Series G3000
  • Accessories are designed to help user adapt their 3M™ Hard Hats to suit individual needs
TRUSTED_SUSTAINABILITY
Specifications
Color Black
For Use With (Equipment) Hard Hat Series G3000
Product Type Chin Strap