missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The GIMAP4 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Linea cellulare | The lysate was created in HEK293T cells, using plasmid ID RC24653 and based on accession number NM_18326. The protein contains a C-terminal DDK tag. |
| Da utilizzare con (applicazione) | Western Blot |
| Quantità | 0.1 mg |
| Tampone di conservazione | RIPA buffer |
| Purezza | Protein |
| Test di specificità | Homo sapiens GTPase, IMAP family member 4 (GIMAP4), mRNA. |
| Content And Storage | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?