Learn More
Hanna Instruments™ Glass Cuvette Sets for Checker HC Colorimeters and Benchtop Photometers
Product Code. 10751571
Change view
Click to view available options
For Use With (Equipment):
HI 83 Series Photometers, HI 83 Series Turbidimeters
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10751571 HI 83 Series Photometers, HI 83 Series Turbidimeters Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10751571 Supplier Hanna Instruments™ Supplier No. HI731321
  • ONLINE EXCLUSIVE PRICE
    66.48€
    31.92€ / pezzi 4
    Risparmia 34.56€
    51% Sconto
Spedizione stimata: 11-12-2026
Replacement optically clear glass cuvettes that are supplied with the Checker HC colorimeters and HI 83 series photometers and turbidimeters.
  • For use with HI 83 series photometers and turbidimeters
  • Replacement glass cuvettes used with the HI7XX Checker HC colorimeters and the HI832XX benchtop photometers
  • Each cuvette has a 10 ml level mark to measure the appropriate volume
  • Index mark for proper alignment in the photometer
  • 10 ml level mark for adding correct volume
  • Optically clear glass
Specifications
For Use With (Equipment) HI 83 Series Photometers, HI 83 Series Turbidimeters