Learn More
Horst™ Glass Fiber GBW Model Fleecy Tape
Product Code. 11903925
Change view
Click to view available options
Material:
Glass fiber
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11903925 Glass fiber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11903925 Supplier Horst™ Supplier No. 100201
  • ONLINE EXCLUSIVE PRICE
    140.90€
    92.42€ / pezzi 1
    Risparmia 48.48€
    34% Sconto
Spedizione stimata: 02-12-2026
Fleecy glass fiber tape for insulating heated sections. Horst™ GBW Glass Fiber Fleecy Tape provides protection up to 450°C.
  • Easy to apply thermal insulation
  • Tape format
  • Easy to remove if necessary
Specifications
Description 3mm thick
Length (Metric) 30 m
Model GBW
Material Glass fiber
Width (Metric) 25mm
Product Type Fleecy Tape
Temperature (Metric) 450°C