Learn More
Hach Lange™ Cella in vetro per campioni
Descrizione
Easy to use and cost effective replacement sample cells for use with 2100N, 2100AN, and lab Ratio turbidimeters are made of glass and contain six sample cell vials with caps.
- For use with Hach Ratio 18900, 2100N and 2100AN Laboratory Turbidmeters
- Replacement sample cells for use with 2100N, 2100AN, and lab Ratio turbidimeters
- For use with Hach Ratio 18900, 2100N and 2100AN laboratory turbidmeters
- Includes six sample cell vials with caps
- Made of glass material
Specifica
Specifica
| Tipo | Glass Sample Cell |
| Da utilizzare con (apparecchiatura) | FC Photoflex/Turb 430 Portable Turbidity Meter and Photometer |
| Materiale | Glass |